Learn More
Medchemexpress LLC Tasisulam | 519055-62-0 | 99.8% | 415.11 | 50 MG
SDP
Catalog No. 5000359665
Encompass_Preferred
Tasisulam is an anticancer agent that induces apoptosis via the intrinsic pathway, leading to cytochrome c release and caspase-dependent cell death. It inhibits mitotic progression and induces vascular normalization.
  • Induces apoptosis via the intrinsic pathway
  • Leads to cytochrome c release and caspase-dependent cell death
  • Inhibits mitotic progression
  • Induces vascular normalization
  • Demonstrates antiproliferative response across tumor histologies
  • Increases 4N DNA and G2-M accumulation
  • Blocks VEGF, epidermal growth factor, and fibroblast growth factor-induced endothelial cell cord formation
Catalog No. 50-003-59665 Supplier Medchemexpress LLC Supplier No. HY1480450MG
Tasisulam is an anticancer agent that induces apoptosis via the intrinsic pathway, leading to cytochrome c release and caspase-dependent cell death. It inhibits mitotic progression and induces vascular normalization.
  • Induces apoptosis via the intrinsic pathway
  • Leads to cytochrome c release and caspase-dependent cell death
  • Inhibits mitotic progression
  • Induces vascular normalization
  • Demonstrates antiproliferative response across tumor histologies
  • Increases 4N DNA and G2-M accumulation
  • Blocks VEGF, epidermal growth factor, and fibroblast growth factor-induced endothelial cell cord formation