Learn More
TARGETMOL CHEMICALS INC TASK-1-IN-1 5MG
SDP
Catalog No. 502579250
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. TASK-1-IN-1 is a potent and selective inhibitor of TASK-1 (Potassium Channel) (IC50 148 nM).TASK-1-IN-1 has a reduced inhibitory effect on the TASK-3 channel with an IC50 of 1750 nM.TASK-1-IN-1 has anticancer effects. purity: 100%
Catalog No. 50-257-9250 Supplier TARGETMOL CHEMICALS INC Supplier No. T720645MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. TASK-1-IN-1 is a potent and selective inhibitor of TASK-1 (Potassium Channel) (IC50 148 nM).TASK-1-IN-1 has a reduced inhibitory effect on the TASK-3 channel with an IC50 of 1750 nM.TASK-1-IN-1 has anticancer effects. purity: 100%