Learn More
Cayman Chemical TASP0415914, Purity: ≥98%, CAS: 1292300-75-4, 10 mg
SDP
Catalog No. 502978562
Encompass_Preferred
A PI3Kγ inhibitor (IC50 = 29 nM); selective for PI3Kγ over Akt (IC50 = 294 nM); reduces ankle redness and swelling in a mouse model of collagen-induced arthritis at 30 and 100 mg/kg
Catalog No. 50-297-8562 Supplier Cayman Chemical Supplier No. 4114510 MG
A PI3Kγ inhibitor (IC50 = 29 nM); selective for PI3Kγ over Akt (IC50 = 294 nM); reduces ankle redness and swelling in a mouse model of collagen-induced arthritis at 30 and 100 mg/kg