Learn More
Medchemexpress LLC HY-P0165 10mg Medchemexpress, Taspoglutide CAS:275371-94-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP016510MG
Encompass_Preferred
Medchemexpress, HY-P0165 10mg Taspoglutide CAS:275371-94-3 Taspoglutide is a long-acting glucagon-like peptide 1 ( GLP-1 ) receptor agonist developed for treatment of type 2 diabetes, with an EC 50 value of 0.06 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4035