Learn More
Cayman Chemical TaspoglutIdeacetate 10mg
SDP
Catalog No. 502944740
Encompass_Preferred
An agonist of GLP-1R (Ki = 1.1 nM for the human receptor); induces cAMP accumulation in CHO-K1 cells expressing human GLP-1R (EC50 = 0.06 nM); decreases blood levels of glucose and increases blood levels of insulin in an oral glucose tolerance test in Zucker diabetic obese rats at 1 mg/animal per week; reduces blood levels of GIP, plasma levels of triglycerides, and body weight in the same model
Catalog No. 50-294-4740 Supplier Cayman Chemical Supplier No. 4125810 MG
An agonist of GLP-1R (Ki = 1.1 nM for the human receptor); induces cAMP accumulation in CHO-K1 cells expressing human GLP-1R (EC50 = 0.06 nM); decreases blood levels of glucose and increases blood levels of insulin in an oral glucose tolerance test in Zucker diabetic obese rats at 1 mg/animal per week; reduces blood levels of GIP, plasma levels of triglycerides, and body weight in the same model