Learn More
Apexbio Technology LLC Tasquinimod, 10mg. Cas: 254964-60-8 MFCD: MFCD18251454. Antiangiogenic and antineoplastic agent
SDP
Supplier:  Apexbio Technology LLC A386010
Encompass_Preferred
Tasquinimod is an orally administered quinoline-3-carboxamide with potent antiangiogenic and antitumorigenic ability that has shown promise in the treatment of advanced prostate cancers [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4844