Learn More
TARGETMOL CHEMICALS INC TASQUINIMOD 10MG
SDP
Catalog No. 502576471
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tasquinimod (ABR-215050) is a quinoline-3-carboxamide linomide analog with antiangiogenic and potential antineoplastic activities. Tasquinimod has been shown to decrease blood vessel density but the exact mechanism of action is not known. This agent has also been shown to augment the antineoplastic effects of docetaxel and androgen ablation in a murine model of prostate cancer involving human prostate cancer xenografts. purity: 99%
Catalog No. 50-257-6471 Supplier TARGETMOL CHEMICALS INC Supplier No. T669510MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tasquinimod (ABR-215050) is a quinoline-3-carboxamide linomide analog with antiangiogenic and potential antineoplastic activities. Tasquinimod has been shown to decrease blood vessel density but the exact mechanism of action is not known. This agent has also been shown to augment the antineoplastic effects of docetaxel and androgen ablation in a murine model of prostate cancer involving human prostate cancer xenografts. purity: 99%