Learn More
Cayman Chemical Tasquinimod, Purity: ≥98%, CAS: 254964-60-8, 10 mg
SDP
Catalog No. 502979866
Encompass_Preferred
An orally-active quinoline-3-carboxamide that inhibits tumor angiogenesis and supplements radiation or chemotherapy in animal models of prostate cancer
Catalog No. 50-297-9866 Supplier Cayman Chemical Supplier No. 1769210 MG
An orally-active quinoline-3-carboxamide that inhibits tumor angiogenesis and supplements radiation or chemotherapy in animal models of prostate cancer