Learn More
Apexbio Technology LLC Tasquinimod 254964-60-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064265
Encompass_Preferred
Tasquinimod (CAS 254964-60-8) is an orally bioavailable quinoline-3-carboxamide compound with antiangiogenic and antitumor properties It modulates angiogenesis primarily through dual inhibition of S100A9/TLR4 signaling in myeloid-derived suppressor cells (MDSCs) and downregulation of HIF-1 and VEGF expression in tumor and endothelial cells It also enhances thrombospondin-1 (TSP-1) expression further suppressing neovascularization Preclinical studies in human prostate cancer xenograft models indicate significant antineoplastic activity particularly in combination with docetaxel radiation therapy or androgen-depletion strategies suggesting its therapeutic potential for castration-resistant prostate cancer (CRPC)
Catalog No. 50-000-64265 Supplier Apexbio Technology LLC Supplier No. A38605.1
Tasquinimod (CAS 254964-60-8) is an orally bioavailable quinoline-3-carboxamide compound with antiangiogenic and antitumor properties It modulates angiogenesis primarily through dual inhibition of S100A9/TLR4 signaling in myeloid-derived suppressor cells (MDSCs) and downregulation of HIF-1 and VEGF expression in tumor and endothelial cells It also enhances thrombospondin-1 (TSP-1) expression further suppressing neovascularization Preclinical studies in human prostate cancer xenograft models indicate significant antineoplastic activity particularly in combination with docetaxel radiation therapy or androgen-depletion strategies suggesting its therapeutic potential for castration-resistant prostate cancer (CRPC)