Learn More
Selleck Chemical LLC Tasquinimod-5mg
SDP
Catalog No. 502389662
Encompass_Preferred
Tasquinimod (ABR-215050) is an orally active antiangiogenic agent by allosterically inhibiting HDAC4 signalling. Phase 3.
Catalog No. 50-238-9662 Supplier Selleck Chemical LLC Supplier No. S76175MG
Tasquinimod (ABR-215050) is an orally active antiangiogenic agent by allosterically inhibiting HDAC4 signalling. Phase 3.