Learn More
Medchemexpress LLC Tasquinimod-d3 | 1416701-99-9 | 99.3% | 409.37 | C20H14D3F3N2O4 | 1 MG
SDP
Catalog No. 5000242375
Encompass_Preferred
Tasquinimod-d3 is a deuterium-labeled isotopologue of tasquinimod supplied as a high-purity reference standard for analytical and research applications. It is intended for use as an internal standard or tracer in mass spectrometry, chromatography, and pharmacokinetic studies; not for human therapeutic use.
  • High isotopic labeling for accurate mass spectrometry quantitation.
  • High chemical purity (99.3%) for reliable analytical results.
  • Available in small, pre-weighed quantities suitable for LC-MS workflows.
  • Characterized molecular weight and formula for confident identification.
  • Provided with documentation to support traceability and analytical use.
Catalog No. 50-002-42375 Supplier Medchemexpress LLC Supplier No. HY10528S1MG
Tasquinimod-d3 is a deuterium-labeled isotopologue of tasquinimod supplied as a high-purity reference standard for analytical and research applications. It is intended for use as an internal standard or tracer in mass spectrometry, chromatography, and pharmacokinetic studies; not for human therapeutic use.
  • High isotopic labeling for accurate mass spectrometry quantitation.
  • High chemical purity (99.3%) for reliable analytical results.
  • Available in small, pre-weighed quantities suitable for LC-MS workflows.
  • Characterized molecular weight and formula for confident identification.
  • Provided with documentation to support traceability and analytical use.