Learn More
Medchemexpress LLC Tasquinimod-d3 5mg | 1416701-99-9 | 5MG
SDP
Catalog No. 5000242376
Encompass_Preferred
Tasquinimod-d3 (ABR-215050-d3) is the deuterium labeled Tasquinimod (HY-10528) Tasquinimod is an oral antiangiogenic agent which plays an important role in castration-resistant prostate cancer Tasquinimod binds to the regulatory Zn2 binding domain of HDAC4 with Kd of 10-30 nM Tasquinimod also is a S100A9 inhibitor[1 [2 [3
Catalog No. 50-002-42376 Supplier Medchemexpress LLC Supplier No. HY10528S5MG
Tasquinimod-d3 (ABR-215050-d3) is the deuterium labeled Tasquinimod (HY-10528) Tasquinimod is an oral antiangiogenic agent which plays an important role in castration-resistant prostate cancer Tasquinimod binds to the regulatory Zn2 binding domain of HDAC4 with Kd of 10-30 nM Tasquinimod also is a S100A9 inhibitor[1 [2 [3