Learn More
Medchemexpress LLC Tasurgratinib succinate | 1879965-80-6 | 99.2% | 705.8 g/mol | C32H37N5O6.3/2C4H6O4 | 10 MG
SDP
Catalog No. 5000247695
Encompass_Preferred
Tasurgratinib succinate (E7090) is an orally available, selective inhibitor of fibroblast growth factor receptors FGFR1-3 used in preclinical oncology research. It potently inhibits FGFR signaling at low-nanomolar concentrations and has demonstrated antiproliferative activity in FGFR-driven cancer cell lines.
  • Selective FGFR1-3 inhibition with low-nanomolar potency.
  • Demonstrated antiproliferative activity in FGFR-driven cell lines.
  • Supplied as the succinate salt for preclinical experiments.
  • High purity suitable for biochemical and cellular assays.
  • Provided in small-mass packages for research use.
Catalog No. 50-002-47695 Supplier Medchemexpress LLC Supplier No. HY101466A10MG
Tasurgratinib succinate (E7090) is an orally available, selective inhibitor of fibroblast growth factor receptors FGFR1-3 used in preclinical oncology research. It potently inhibits FGFR signaling at low-nanomolar concentrations and has demonstrated antiproliferative activity in FGFR-driven cancer cell lines.
  • Selective FGFR1-3 inhibition with low-nanomolar potency.
  • Demonstrated antiproliferative activity in FGFR-driven cell lines.
  • Supplied as the succinate salt for preclinical experiments.
  • High purity suitable for biochemical and cellular assays.
  • Provided in small-mass packages for research use.