Learn More
Medchemexpress LLC Tasurgratinib succinate | 1879965-80-6 | 99.2% | 764.8 g/mol | C32H37N5O6·3/2C4H6O4 | 50 MG
SDP
Catalog No. 5000246651
Encompass_Preferred
E7090 succinate (tasurgratinib succinate) is the succinate salt of tasurgratinib, an orally active small-molecule inhibitor selective for fibroblast growth factor receptors FGFR1-3. It is supplied as a high-purity research compound with documented solubility and in vivo formulation protocols for preclinical studies.
Catalog No. 50-002-46651 Supplier Medchemexpress LLC Supplier No. HY101466A50MG
E7090 succinate (tasurgratinib succinate) is the succinate salt of tasurgratinib, an orally active small-molecule inhibitor selective for fibroblast growth factor receptors FGFR1-3. It is supplied as a high-purity research compound with documented solubility and in vivo formulation protocols for preclinical studies.