Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tasurgratinib succinate | 1879965-80-6 | 99.2% | 764.82 g/mol | C32H37N5O6·3/2 C4H6O4 | 25 MG
SDP

Catalog No. 5000248245
Encompass_Preferred

Tasurgratinib succinate is an orally available, selective small-molecule inhibitor of fibroblast growth factor receptors FGFR1, FGFR2, and FGFR3. Supplied as the succinate salt for research use, the compound is characterized by a defined molecular formula and high purity for preclinical and translational studies probing FGFR-driven signaling and anticancer activity.

  • Selective FGFR1-3 inhibition with low nanomolar potency.
  • High reported purity suitable for research applications.
  • Supplied as a succinate salt for improved stability and handling.
  • Molecular weight and formula provided for analytical characterization.
  • Soluble in common organic solvents for in vitro testing.

Catalog No. 50-002-48245 Supplier Medchemexpress LLC Supplier No. HY101466A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tasurgratinib succinate is an orally available, selective small-molecule inhibitor of fibroblast growth factor receptors FGFR1, FGFR2, and FGFR3. Supplied as the succinate salt for research use, the compound is characterized by a defined molecular formula and high purity for preclinical and translational studies probing FGFR-driven signaling and anticancer activity.

  • Selective FGFR1-3 inhibition with low nanomolar potency.
  • High reported purity suitable for research applications.
  • Supplied as a succinate salt for improved stability and handling.
  • Molecular weight and formula provided for analytical characterization.
  • Soluble in common organic solvents for in vitro testing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.