Learn More
Medchemexpress LLC Tasurgratinib succinate | 1879965-80-6 | 99.2% | 764.82 g·mol⁻¹ | C32H37N5O6 · 3/2 C4H6O4 | 5 MG
SDP
Catalog No. 5000247071
Encompass_Preferred
Tasurgratinib succinate is the succinate salt of an orally active, selective inhibitor of FGFR1-3 used in preclinical and research applications. The compound shows low-nanomolar potency against FGFR1, FGFR2, and FGFR3 and is supplied as a high-purity solid or as a DMSO solution for laboratory studies.
  • High purity suitable for research use.
  • Potent FGFR1/2/3 inhibition at low nanomolar concentrations.
  • Available as solid and as DMSO solution for assay flexibility.
  • Soluble in DMSO at high concentration (≈100 mg/mL).
  • Pack sizes appropriate for preclinical experiments.
Catalog No. 50-002-47071 Supplier Medchemexpress LLC Supplier No. HY101466A5MG
Tasurgratinib succinate is the succinate salt of an orally active, selective inhibitor of FGFR1-3 used in preclinical and research applications. The compound shows low-nanomolar potency against FGFR1, FGFR2, and FGFR3 and is supplied as a high-purity solid or as a DMSO solution for laboratory studies.
  • High purity suitable for research use.
  • Potent FGFR1/2/3 inhibition at low nanomolar concentrations.
  • Available as solid and as DMSO solution for assay flexibility.
  • Soluble in DMSO at high concentration (≈100 mg/mL).
  • Pack sizes appropriate for preclinical experiments.