Learn More
Fujifilm Medical Systems μTASWako AFP-L3 Sample Dilution Buffer
Catalog No. 23222004 Shop All FujiFilm Products
Change view
Click to view available options
Quantity:
3 x 10 mL
Catalog No. Quantity
23-222-004 3 x 10 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-222-004 Supplier Fujifilm Medical Systems Supplier No. 99761501
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with the μTASWako i30 Immunoanalyzer System
  • Clinical Diagnostics Protein and Cell Analysis μTASWako AFP-L3 Dilution Buffer
  • When AFP levels are too high dilute the sample with AFP-L3 dilution buffer
  • 3 x 10 mL/Ea.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) μTASWako i30 Immunoanalyzer System
Quantity 3 x 10 mL