Learn More
Fujifilm Medical Systems μTASWako AFP-L3
Catalog No. 23222000 Shop All FujiFilm Products
Change view
Click to view available options
Quantity:
100 Tests
Catalog No. Quantity
23-222-000 100 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-222-000 Supplier Fujifilm Medical Systems Supplier No. 99960601
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
For use with the μTASWako i30 Immunoanalyzer System
  • Clinical Diagnostics Protein and Cell Analysis μTASWako AFP-L3 Quantitatively measure lectin-reactive AFP (AFP-L3) in human serum
  • Reportable range of 0.5% to 99.5%
  • For in vitro diagnostic use to aid in HCC risk assessment
  • 100 tests/Ea.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) μTASWako i30 Immunoanalyzer System
Quantity 100 Tests