Learn More
Fujifilm Medical Systems μTASWako DCP Calibrator Set
Catalog No. 23222006 Shop All FujiFilm Products
Change view
Click to view available options
Quantity:
3 x 2 mL
Catalog No. Quantity
23-222-006 3 x 2 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-222-006 Supplier Fujifilm Medical Systems Supplier No. 99961201
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with the μTASWako i30 Immunoanalyzer System
  • Clinical Diagnostics Protein and Cell Analysis μTASWako DCP Calibrator Set Quality Control
  • Designed for use with the μTASWako DCP Test System
  • For in vitro diagnostic use to aid in HCC risk assessment
  • 1 Set/Ea.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) μTASWako i30 Immunoanalyzer System
Quantity 3 x 2 mL