Learn More
Fujifilm Medical Systems μTASWako DCP
Catalog No. 23222005 Shop All FujiFilm Products
Change view
Click to view available options
Quantity:
100 Tests
Catalog No. Quantity
23-222-005 100 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-222-005 Supplier Fujifilm Medical Systems Supplier No. 99560701
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
For use with the μTASWako i30 Immunoanalyzer System
  • Clinical Diagnostics Protein and Cell Analysis μTASWako DCP Quantitatively measure des-gamma-carboxy prothrombin (DCP) in human serum
  • For in vitro diagnostic use to aid in HCC risk assessment
  • 100 tests/Ea.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) μTASWako i30 Immunoanalyzer System
Quantity 100 Tests