Learn More
Fujifilm Medical Systems μTASWako i30 Immunoanalyzer
Catalog No. 23222012 Shop All FujiFilm Products
Change view
Click to view available options
For Use With (Application):
Aid in Liver Cancer Risk Assessment
Catalog No. For Use With (Application)
23-222-012 Aid in Liver Cancer Risk Assessment
1 options
This item is not returnable. View return policy
Catalog No. 23-222-012 Supplier Fujifilm Medical Systems Supplier No. 99160301
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
μTASWako i30 Immunoanalyzer System to aid in Liver Cancer Risk Assessment
  • The automated μTASWako i30 is a microfluidic-based clinical immunoanalyzer that minimizes instrument hands-on time while maximizing efficiency through integration and automation of clinical laboratory processes
  • The μTASWako AFP-L3 and DCP tests are designed for use with the μTASWako i30 system to aid in hepatocellular carcinoma (HCC) risk assessment.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Aid in Liver Cancer Risk Assessment