Learn More
Medchemexpress LLC TAT-14 | 1362661-34-4 | 99.7% | 3173.59 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP13281MG
Encompass_Preferred
TAT-14 is a 14-mer peptide that functions as an Nrf2 activator with anti-inflammatory properties. It does not affect Nrf2 mRNA expression, but instead increases Nrf2 protein levels by targeting the Nrf2 binding site on Keap1.
  • Functions as an Nrf2 activator
  • Has anti-inflammatory properties
  • Increases Nrf2 protein levels by targeting the Nrf2 binding site on Keap1
  • Soluble in H2O at 100 mg/mL (31.51 mM); requires ultrasonic
  • Soluble in PBS at 50 mg/mL (15.76 mM); clear solution; requires ultrasonic
  • For research use only
Catalog No. 50-004-24101