Learn More
CPC Scientific H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Gly-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Gly-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific TAT003A
Encompass
ONE-LETTER SEQUENCE: YGRKKRRQRRRGYGRKKRRQRRRGMOLECULAR FORMULA: C132H240N66O29MOLECULAR WEIGHT:3215.79STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1159916-66-1], RESEARCH AREA: Antimicrobial
Catalog No. 50-211-0541