Learn More
Carbosynth LLC. TAT (47-57), 1mg, MW: 1559.86, Molecular formula: C64H118N32O14, CAS# 191936-91-1
SDP
Supplier:  Carbosynth LLC. TAT3768PI1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-OH; HIV TAT (47-57), H-YGRKKRRQRRR-OH, Application: TAT(Transactivated-transcripiton) Peptide Derived from the HIV TAT protein; is a Cell-Penetrating Peptide, Salt Form:, Trifluroacetate Salt, Storage Temperature: -20C, References: H. Wang, et al., J. Clin. Invest., 109, 1463 (2002), Ziegler, et al., Biochem., 44, 138 (2005), P.A. Wender, et al., PNAS, 97, 13003 (2000)
Catalog No. 50-168-7194