Learn More
Novus Biologicals™ Tat-Beclin 1 L11 Autophagy Inducing Peptide

Specifications
Specifications
| Species | Non-species Specific |
| Content And Storage | Store at -20°C in powder form. Store at -80°C once reconstituted. |
| Gene Alias | Autophagy Inducing peptide, Tat-Beclin, Tat-Beclin 1, Tat-Beclin 1 peptide, Tat-Beclin peptide, Tat-L11 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 3.08 kDa |
| Quantity | 0.1 mg |
| Research Category | Autophagy |
| Storage Buffer | This product is supplied lyophilized. Purity is ≥ to 97% (HPLC) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.