Learn More
Selleck Chemical LLC Tat-beclin 1 (Tat-BECN1) 2mg 1423821-88-8
SDP
Supplier:  Selleck Chemical LLC S85952MG
Encompass_Preferred
Tat-beclin 1 (Tat-BECN1), a peptide known to stimulate autophagy through mobilization of endogenous Beclin 1, induces autophagy in vitro and in vivo and improves clinical outcomes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8133