Learn More
General Laboratory Products TAT Broth 490 mL, Sterility Testing, Microbiological Examination
Catalog No. NC2072164
Encompass_Preferred
General Laboratory Products TAT Broth is a specialized liquid microbiological medium for detecting and cultivating microorganisms in cosmetics and topical drugs. Its enriched formula neutralizes common preservatives, ensuring precise sterility testing. Suitable for quality control in the pharmaceutical and cosmetic industries, it supports the growth of a wide variety of microorganisms, delivering reliable results for testing viscous materials.
Catalog No. NC2072164 Supplier General Laboratory Products Supplier No. 32WM3600490
General Laboratory Products TAT Broth is a specialized liquid microbiological medium for detecting and cultivating microorganisms in cosmetics and topical drugs. Its enriched formula neutralizes common preservatives, ensuring precise sterility testing. Suitable for quality control in the pharmaceutical and cosmetic industries, it supports the growth of a wide variety of microorganisms, delivering reliable results for testing viscous materials.