Learn More
Medchemexpress LLC TAT-CBD3A6K acetate | 99.8% | 3249.83 (free base) | 1 MG
SDP
Catalog No. 5000425094
Encompass_Preferred
TAT-CBD3A6K acetate is a modified TAT-CBD3 peptide that reduces T- and R-type voltage-dependent calcium currents in dorsal root ganglion (DRG) neurons. It shows anti-nociceptive effects in a model of AIDS-induced peripheral neuropathy by preventing CRMP-2-mediated enhancement of T- and R-type calcium channel function.
  • Peptides and derivatives
  • Inhibitors and substrates
  • Neurological applications
  • Eye or ear disease research
  • Inhibits calcium channels
  • Used in AIDS research
  • Anti-nociceptive effects
  • Peripheral neuropathy studies
  • DRG studies
Catalog No. 50-004-25094 Supplier Medchemexpress LLC Supplier No. HYP10358A1MG
TAT-CBD3A6K acetate is a modified TAT-CBD3 peptide that reduces T- and R-type voltage-dependent calcium currents in dorsal root ganglion (DRG) neurons. It shows anti-nociceptive effects in a model of AIDS-induced peripheral neuropathy by preventing CRMP-2-mediated enhancement of T- and R-type calcium channel function.
  • Peptides and derivatives
  • Inhibitors and substrates
  • Neurological applications
  • Eye or ear disease research
  • Inhibits calcium channels
  • Used in AIDS research
  • Anti-nociceptive effects
  • Peripheral neuropathy studies
  • DRG studies