Learn More
Medchemexpress LLC Tat-cbd3a6k (acetate) | 99.8% | 3249.83 (free base) | 5 MG
SDP
Catalog No. 5000425151
Encompass_Preferred
This modified peptide reduces T- and R-type voltage-dependent calcium currents in dorsal root ganglion (DRG) neurons. It demonstrates anti-nociceptive effects in a model of AIDS-induced peripheral neuropathy. This action is achieved by preventing enhancement of T- and R-type calcium channel function.
  • Reduces T- and R-type voltage-dependent calcium currents.
  • Shows anti-nociceptive effects.
  • Acts by preventing enhancement of calcium channel function.
Catalog No. 50-004-25151 Supplier Medchemexpress LLC Supplier No. HYP10358A5MG
This modified peptide reduces T- and R-type voltage-dependent calcium currents in dorsal root ganglion (DRG) neurons. It demonstrates anti-nociceptive effects in a model of AIDS-induced peripheral neuropathy. This action is achieved by preventing enhancement of T- and R-type calcium channel function.
  • Reduces T- and R-type voltage-dependent calcium currents.
  • Shows anti-nociceptive effects.
  • Acts by preventing enhancement of calcium channel function.