Learn More
Medchemexpress LLC Tat-NR2B9c TFA | 1834571-04-8 | 99.7% | 2632.90 | 25 MG
SDP
Catalog No. 5000426467
Encompass_Preferred
Tat-NR2B9c TFA is a potent inhibitor of postsynaptic density-95 (PSD-95). It selectively disrupts the interaction between PSD-95 and NMDAR, inhibiting the binding of NR2A and NR2B to PSD-95. This compound also inhibits the interaction between neuronal nitric oxide synthase (nNOS) and PSD-95, demonstrating significant neuroprotective efficacy.
  • Potent PSD-95 inhibitor with high affinity for PSD-95d2 (EC50 6.7 nM)
  • Inhibits NMDAR2A, NMDAR2B, and NMDAR2C binding to PSD-95
  • Blocks interaction between PSD-95 and nNOS
  • Reduces infarction volume in vivo
Catalog No. 50-004-26467 Supplier Medchemexpress LLC Supplier No. 25MGHYP0117A
Tat-NR2B9c TFA is a potent inhibitor of postsynaptic density-95 (PSD-95). It selectively disrupts the interaction between PSD-95 and NMDAR, inhibiting the binding of NR2A and NR2B to PSD-95. This compound also inhibits the interaction between neuronal nitric oxide synthase (nNOS) and PSD-95, demonstrating significant neuroprotective efficacy.
  • Potent PSD-95 inhibitor with high affinity for PSD-95d2 (EC50 6.7 nM)
  • Inhibits NMDAR2A, NMDAR2B, and NMDAR2C binding to PSD-95
  • Blocks interaction between PSD-95 and nNOS
  • Reduces infarction volume in vivo