Learn More
Medchemexpress LLC Tat-NR2Baa | 847829-41-8 | 96.9% | 2474.83 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP23071MG
Encompass_Preferred
This product is a control peptide for another peptide, though it is inactive. Its sequence is similar to the active peptide, but a double-point mutation in its COOH terminal tSXV motif renders it unable to bind to PSD-95. The active peptide is a membrane-permeant peptide that interferes with PSD-95/NMDAR binding, leading to the uncoupling of NR2B- and/or NR2A-type NMDARs from PSD-95.
  • Functions as a control peptide for an active counterpart.
  • Inactive, useful for comparative studies.
  • Features a double-point mutation in the COOH terminal tSXV motif.
  • Incapable of binding PSD-95.
  • The active version of this peptide disrupts PSD-95/NMDAR binding.
  • The active version uncouples NR2B- and/or NR2A-type NMDARs from PSD-95.
Catalog No. 50-004-23392