Learn More
Medchemexpress LLC Tat-peptide 190-208 TFA | 98.4% | 3393.45 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP5118A500UG
Encompass_Preferred
Tat-peptide 190-208 TFA is a cell-permeable and Tat-labeled fusion peptide, corresponding to residues 190-208 of rat G3BP1. The Tat sequence from HIV is placed at the least conserved end for cell permeability. This peptide increases axon growth and the number of neurites per neuron, likely through an axon intrinsic mechanism. It can also be used for ischemic protection during endovascular repair for intracranial aneurysms. This product is for research use only.
  • Cell-permeable and Tat-labeled fusion peptide
  • Corresponds to residues 190-208 of rat G3BP1
  • Increases axon growth and number of neurites per neuron
  • Used for ischemic protection during endovascular repair for intracranial aneurysms
Catalog No. 50-003-99151