Learn More
Medchemexpress LLC TAT peptide TFA | 99.8% | 1735.93 | 25 MG
SDP
Catalog No. 5000426472
Encompass_Preferred
TAT peptide (TFA) is a cell penetrating peptide (GRKKRRQRRRPQ) derived from the trans-activating transcriptional activator (Tat) from HIV-1. It has been studied in functionalized hybrid nanoparticles for magnetic enrichment and optical detection in cell separation and rapid cell labelling, demonstrating good biocompatibility.
  • Derived from trans-activating transcriptional activator (Tat) from HIV-1
  • Cell penetrating peptide (GRKKRRQRRRPQ)
  • Used in functionalized hybrid nanoparticles for cell separation
  • Used in rapid cell labelling
  • Shows good biocompatibility
  • For research use only
Catalog No. 50-004-26472 Supplier Medchemexpress LLC Supplier No. 25MGHYP0282A
TAT peptide (TFA) is a cell penetrating peptide (GRKKRRQRRRPQ) derived from the trans-activating transcriptional activator (Tat) from HIV-1. It has been studied in functionalized hybrid nanoparticles for magnetic enrichment and optical detection in cell separation and rapid cell labelling, demonstrating good biocompatibility.
  • Derived from trans-activating transcriptional activator (Tat) from HIV-1
  • Cell penetrating peptide (GRKKRRQRRRPQ)
  • Used in functionalized hybrid nanoparticles for cell separation
  • Used in rapid cell labelling
  • Shows good biocompatibility
  • For research use only