Learn More
Medchemexpress LLC TAT-QFNP12 acetate | 3006.47 (free base) | 1 MG
SDP
Catalog No. 5000425393
Encompass_Preferred
TAT-QFNP12 acetate is a peptide that plays a crucial role in cellular signaling pathways. It functions by blocking the NDRG2-PPM1A binding, which in turn reduces Smad2/3 phosphorylation. This peptide is also known to decrease astrocytic MMP-9 production and mitigate blood-brain barrier (BBB) disruption following subarachnoid hemorrhage (SAH).
  • Blocks NDRG2-PPM1A binding
  • Reduces Smad2/3 phosphorylation
  • Decreases astrocytic MMP-9 production
  • Mitigates blood-brain barrier (BBB) disruption after subarachnoid hemorrhage (SAH)
Catalog No. 50-004-25393 Supplier Medchemexpress LLC Supplier No. HYP10359A1MG
TAT-QFNP12 acetate is a peptide that plays a crucial role in cellular signaling pathways. It functions by blocking the NDRG2-PPM1A binding, which in turn reduces Smad2/3 phosphorylation. This peptide is also known to decrease astrocytic MMP-9 production and mitigate blood-brain barrier (BBB) disruption following subarachnoid hemorrhage (SAH).
  • Blocks NDRG2-PPM1A binding
  • Reduces Smad2/3 phosphorylation
  • Decreases astrocytic MMP-9 production
  • Mitigates blood-brain barrier (BBB) disruption after subarachnoid hemorrhage (SAH)