Learn More
Medchemexpress LLC HY-P0281A 1mg Medchemexpress, TAT (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0281A1MG
Encompass_Preferred
Medchemexpress, HY-P0281A 1mg TAT (TFA) CAS: TAT TFA (YGRKKRRQRRR) is derived from the transactivator of transcription (TAT) of human immunodeficiency virus (HIV-1) and is a cell-penetrating peptide. TAT can increase the yields and the solubility of heterologous proteins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0523