Learn More
Cayman Chemical TatCBD3A6KtrIfluoroACTAte 10mg
SDP
Catalog No. 502918192
Encompass_Preferred
A peptide; prevents decreases in the paw withdrawal threshold in a rat model of stavudine-induced antiretroviral neuropathic pain at 10 mg/kg and decreases the number of action potentials from DRG neurons isolated from the same rats; dural administration decreases capsaicin-induced meningeal blood flow in rats
Catalog No. 50-291-8192 Supplier Cayman Chemical Supplier No. 3744910 MG
A peptide; prevents decreases in the paw withdrawal threshold in a rat model of stavudine-induced antiretroviral neuropathic pain at 10 mg/kg and decreases the number of action potentials from DRG neurons isolated from the same rats; dural administration decreases capsaicin-induced meningeal blood flow in rats