Learn More
Cayman Chemical TatCIRPtrIfluoroACTAte sa 10mg
SDP
Catalog No. 502918190
Encompass_Preferred
A peptide inhibitor of the MD-2-CIRP protein-protein interaction; binds to MD-2 and disrupts the interaction between MD-2 and CIRP in co-immunoprecipitation assays; reduces infarct volume in a mouse model of MCAO-induced stroke at 10 and 20 mg/kg; reduces infarct volume and decreases the time to pick up and withdraw food with the stroke-affected arm in a rhesus monkey model of thrombosis-induced stroke
Catalog No. 50-291-8190 Supplier Cayman Chemical Supplier No. 3744610 MG
A peptide inhibitor of the MD-2-CIRP protein-protein interaction; binds to MD-2 and disrupts the interaction between MD-2 and CIRP in co-immunoprecipitation assays; reduces infarct volume in a mouse model of MCAO-induced stroke at 10 and 20 mg/kg; reduces infarct volume and decreases the time to pick up and withdraw food with the stroke-affected arm in a rhesus monkey model of thrombosis-induced stroke