Learn More
Enterprise Technology Solutions Tatco Security Seal, 3/10"Wx13-1/2"Lx1/100"H, 100/PK, Blue
SDP
Catalog No. 5000739662
Encompass_Preferred
Security seals feature an adjustable construction for use on a wide variety of applications. 112 lb. break strength stays on tightly and prevents it from snapping under pressure. Sequentially numbered seals help you easily track every item. Plus, its easy-to-use design allows you to quickly seal items to prevent dislocating.
Catalog No. 50-007-39662 Supplier Enterprise Technology Solutions Supplier No. TCO26300
Security seals feature an adjustable construction for use on a wide variety of applications. 112 lb. break strength stays on tightly and prevents it from snapping under pressure. Sequentially numbered seals help you easily track every item. Plus, its easy-to-use design allows you to quickly seal items to prevent dislocating.