Learn More
Cayman Chemical Tat-M2NX (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502918188
Encompass_Preferred
A peptide TRPM2 antagonist; inhibits hydrogen peroxide-induced calcium influx in HEK293 cells expressing human TRPM2 from 25-100 µM; reduces brain infarct volume in male mice in a model of MCAO-induced stroke at 20 mg/kg pre- or 3-hours-post occlusion
Catalog No. 50-291-8188 Supplier Cayman Chemical Supplier No. 3744710 MG
A peptide TRPM2 antagonist; inhibits hydrogen peroxide-induced calcium influx in HEK293 cells expressing human TRPM2 from 25-100 µM; reduces brain infarct volume in male mice in a model of MCAO-induced stroke at 20 mg/kg pre- or 3-hours-post occlusion