Learn More
Cayman Chemical TatNTS PeptIDtrIfluoROC 10mg
SDP
Catalog No. 502918026
Encompass_Preferred
A cell-penetrating peptide; blocks the protein-protein interaction between annexin A1 and importin β, preventing nuclear translocation of annexin A1 in primary mouse hippocampal neurons; reduces infarct size and neuronal apoptosis, as well as decreases the time to reach the platform in the Morris water maze test in a mouse model of MCAO-induced ischemia-reperfusion injury
Catalog No. 50-291-8026 Supplier Cayman Chemical Supplier No. 3744310 MG
A cell-penetrating peptide; blocks the protein-protein interaction between annexin A1 and importin β, preventing nuclear translocation of annexin A1 in primary mouse hippocampal neurons; reduces infarct size and neuronal apoptosis, as well as decreases the time to reach the platform in the Morris water maze test in a mouse model of MCAO-induced ischemia-reperfusion injury