Learn More
Cayman Chemical TatQFNP12trIfluoroacetAT 10mg
SDP
Catalog No. 502919032
Encompass_Preferred
A peptide inhibitor of the NDRG2-PPM1A protein-protein interaction; decreases MMP-9-induced loss of blood-brain barrier endothelial tight junctions, reduces cerebral edema, as well as increases spontaneous activity and symmetry of limb movement, in an endovascular puncture-induced mouse model of subarachnoid hemorrhage at 20 mg/kg
Catalog No. 50-291-9032 Supplier Cayman Chemical Supplier No. 3884510 MG
A peptide inhibitor of the NDRG2-PPM1A protein-protein interaction; decreases MMP-9-induced loss of blood-brain barrier endothelial tight junctions, reduces cerebral edema, as well as increases spontaneous activity and symmetry of limb movement, in an endovascular puncture-induced mouse model of subarachnoid hemorrhage at 20 mg/kg