Learn More
Sino Biological Recombinant Human Tau Protein (Tau-441, AVI Tag), Biotinylated
Catalog No. 502567655
Encompass_Preferred
Recombinant human Tau-441 (216-391) was expressed in E. coli cells with an N-terminal Avi tag and biotinylated in vitro prior to the final chromatography purification.
Catalog No. 50-256-7655 Supplier Sino Biological Supplier No. T0855LNB20
Recombinant human Tau-441 (216-391) was expressed in E. coli cells with an N-terminal Avi tag and biotinylated in vitro prior to the final chromatography purification.