Learn More
Enzo Life Sciences Tau 441 (2N4R variant) (human), (recombinant) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3210100
Encompass_Preferred
Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized from 50mM MES, pH 6.8, 100mM sodium chloride and 0.5mM EGTA. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-1980