Learn More
Sino Biological Recombinant Human Tau Protein (Tau-410)
Catalog No. 502572908
Encompass_Preferred
Recombinant full length human tag-free Tau-410 was expressed in E. coli cells.
Catalog No. 50-257-2908 Supplier Sino Biological Supplier No. T0654N1000
Recombinant full length human tag-free Tau-410 was expressed in E. coli cells.