Learn More
ACROBiosystems ACROBiosystems Human Tau-441 / 2N4R Protein, His Tag
Supplier:  ACROBiosystems TAUH51H31MG
Encompass_Preferred
Recombinant Protein;1MG;Human Tau-441, His Tag (TAU-H51H3) is expressed from E.coli cells. It contains AA Met 1 - Leu 441 (Accession # P10636-8).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 47.9 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7027