Learn More
Sino Biological Recombinant Human Tau Protein (Tau-441), DYRK1A-phosphorylated
Catalog No. 502572877
Encompass_Preferred
Recombinant human tag-free Tau-441 was co-expressed with DYRK1A in E. coli cells. The Tau-441 was phosphorylated by DYRK1A in vivo and in vitro prior to the final chromatography purification.
Catalog No. 50-257-2877 Supplier Sino Biological Supplier No. T0850RN100
Recombinant human tag-free Tau-441 was co-expressed with DYRK1A in E. coli cells. The Tau-441 was phosphorylated by DYRK1A in vivo and in vitro prior to the final chromatography purification.