Learn More
Sino Biological Recombinant Human Tau Protein (Tau-441 (S305N))
Catalog No. 502572800
Encompass_Preferred
Recombinant human tag-free Tau-441 (S305N) was expressed in E. coli cells.
Catalog No. 50-257-2800 Supplier Sino Biological Supplier No. T0856HN1000
Recombinant human tag-free Tau-441 (S305N) was expressed in E. coli cells.