Learn More
Sino Biological Recombinant Human Tau Protein (Tau-441), CAMK2-phosphorylated
Catalog No. 502572888
Encompass_Preferred
Recombinant human tag-free Tau-441 was co-expressed with CAMK2 in E. coli cells. The Tau-441 protein was phosphorylated by CAMK2 in vivo and in vitro prior to the final chromatography purification.
Catalog No. 50-257-2888 Supplier Sino Biological Supplier No. T0850CN1000
Add to Cart
Add to Cart
Request a Quote
Recombinant human tag-free Tau-441 was co-expressed with CAMK2 in E. coli cells. The Tau-441 protein was phosphorylated by CAMK2 in vivo and in vitro prior to the final chromatography purification.