Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tau-D/0N4R Human 1 1mg
SDP

Catalog No. 5000196113
Encompass_Preferred

Tau-D/0N4R Protein, Human (133 a.a., His) is a recombinant fragment of human Tau (MAPT) representing the 0N4R isoform. The protein is expressed in E. coli, purified, and supplied as a lyophilized His-tagged material intended for research applications in Tau biology, microtubule studies, and neurodegeneration research.

  • Recombinant human Tau 0N4R isoform, 133 amino acids.
  • C-terminal 6xHis tag for detection and purification.
  • Expressed in E. coli and supplied as a lyophilized powder.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) for sensitive assays.
  • Stable at -20°C for long-term storage; reconstituted stability at 4°C for one week.

Catalog No. 50-001-96113 Supplier Medchemexpress LLC Supplier No. HYP711021MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tau-D/0N4R Protein, Human (133 a.a., His) is a recombinant fragment of human Tau (MAPT) representing the 0N4R isoform. The protein is expressed in E. coli, purified, and supplied as a lyophilized His-tagged material intended for research applications in Tau biology, microtubule studies, and neurodegeneration research.

  • Recombinant human Tau 0N4R isoform, 133 amino acids.
  • C-terminal 6xHis tag for detection and purification.
  • Expressed in E. coli and supplied as a lyophilized powder.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) for sensitive assays.
  • Stable at -20°C for long-term storage; reconstituted stability at 4°C for one week.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.