Learn More
Medchemexpress LLC Tau-D/0N4R Human 1 500ug
SDP
Catalog No. 5000196606
Encompass_Preferred
Recombinant human Tau-D/0N4R is a 133-amino-acid fragment of microtubule-associated protein tau supplied with a C-terminal 6xHis tag. Expressed in E. coli and provided lyophilized from PBS with 1 mM PMSF, the product is >95% pure by reducing SDS-PAGE and is suitable for biochemical, biophysical, and assay development studies related to tau function and aggregation.
  • Contains 133 amino acids with a C-terminal 6xHis tag.
  • Expressed in E. coli for consistent recombinant production.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Lyophilized from PBS with 1 mM PMSF for stability.
  • Stable refrigerated short term or frozen for long-term storage.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.
Catalog No. 50-001-96606 Supplier Medchemexpress LLC Supplier No. HYP71102500UG
Recombinant human Tau-D/0N4R is a 133-amino-acid fragment of microtubule-associated protein tau supplied with a C-terminal 6xHis tag. Expressed in E. coli and provided lyophilized from PBS with 1 mM PMSF, the product is >95% pure by reducing SDS-PAGE and is suitable for biochemical, biophysical, and assay development studies related to tau function and aggregation.
  • Contains 133 amino acids with a C-terminal 6xHis tag.
  • Expressed in E. coli for consistent recombinant production.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Lyophilized from PBS with 1 mM PMSF for stability.
  • Stable refrigerated short term or frozen for long-term storage.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.